Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc4a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271544.1 | NP_001258473.1 | 4 - 5 | 573 | 87 | |
NM_001271545.1 | NP_001258474.1 | 4 - 5 | 573 | 87 | ||
NM_001271546.1 | NP_001258475.1 | 4 - 5 | 573 | 87 | ||
NM_001271547.1 | NP_001258476.1 | 4 - 5 | 573 | 87 | ||
NM_172830.2 | NP_766418.1 | 4 - 5 | 573 | 87 | ||
GenBank mRNA | AK050187.1 | - | 2 - 3 | 149 | 87 | |
AK085269.1 | - | 4 - 5 | 573 | 87 | ||
AK085424.1 | - | 4 - 5 | 576 | 87 | ||
AK128930.1 | - | 4 - 5 | 575 | 87 | ||
AK142545.1 | - | 4 - 5 | 575 | 87 | ||
AK143939.1 | - | 4 - 5 | 579 | 87 | ||
BC113179.1 | - | 4 - 5 | 568 | 40060674 | 87 | |
BC113785.1 | - | 4 - 5 | 568 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |