Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lax1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159649.1 | NP_001153121.1 | 1 - 2 | 299 | 67 | |
GenBank mRNA | AK040834.1 | - | 1 - 2 | 109 | 67 | |
AK088503.1 | - | 1 - 2 | 52 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Inventoried Probe spans exons |