Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tor1aip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172843.4 | NP_766431.3 | 3 - 4 | 829 | 77 | |
GenBank mRNA | AK038024.1 | - | 3 - 4 | 1091 | 77 | |
CF164425.1 | - | 2 - 3 | 413 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |