Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankmy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172850.3 | NP_766438.2 | 11 - 12 | 2298 | 59 | |
GenBank mRNA | AK029700.1 | - | 11 - 12 | 2300 | 59 | |
BC120887.1 | - | 10 - 11 | 2134 | 40060916 | 59 | |
BC120888.1 | - | 10 - 11 | 2134 | 40060917 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |