Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172855.3 | NP_766443.2 | 3 - 4 | 1947 | 100 | |
GenBank mRNA | AK029323.1 | - | 2219 | 100 | ||
BC116331.1 | - | 3 - 4 | 1946 | 100 | ||
BC116332.1 | - | 3 - 4 | 1947 | 40096906 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |