Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frmd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163732.1 | NP_001157204.1 | 2 - 3 | 423 | 93 | |
NM_172869.4 | NP_766457.1 | 2 - 3 | 423 | 93 | ||
GenBank mRNA | AK034956.1 | - | 2 - 3 | 398 | 93 | |
AK048561.1 | - | 2 - 3 | 366 | 93 | ||
AK079349.1 | - | 2 - 3 | 424 | 93 | ||
BC037727.1 | - | 2 - 3 | 594 | 93 | ||
BC112419.1 | - | 2 - 3 | 331 | 40061426 | 93 | |
BC113795.1 | - | 2 - 3 | 331 | 40061428 | 93 | |
BC118964.2 | - | 2 - 3 | 333 | 40097059 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |