Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400024048.mm.1 |
TC0400002962.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400024048.mm.1 |
NM_172870.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024048.mm.1 |
NM_172870.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024047.mm.1 |
TC0400002962.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400024047.mm.1 |
NM_172870.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024047.mm.1 |
NM_172870.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024045.mm.1 |
TC0400002962.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400024045.mm.1 |
NM_172870.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024045.mm.1 |
NM_172870.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024040.mm.1 |
TC0400002962.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400024040.mm.1 |
NM_172870.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024040.mm.1 |
NM_172870.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024042.mm.1 |
TC0400002962.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400024042.mm.1 |
NM_172870.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024042.mm.1 |
NM_172870.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024043.mm.1 |
TC0400002962.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400024043.mm.1 |
NM_172870.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024043.mm.1 |
NM_172870.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024044.mm.1 |
TC0400002962.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400024044.mm.1 |
NM_172870.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400024044.mm.1 |
NM_172870.4 |
Refseq |