Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Styk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172891.2 | NP_766479.1 | 1 - 2 | 80 | ||
GenBank mRNA | AK033606.1 | - | 1 - 2 | 80 | ||
AK035247.1 | - | 2 - 3 | 80 | |||
AY563053.1 | - | 1 - 2 | 60 | 80 | ||
BC150724.1 | - | 1 - 2 | 9087635 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |