Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Styk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172891.2 | NP_766479.1 | 4 - 5 | 1492 | 59 | |
GenBank mRNA | AK033606.1 | - | 4 - 5 | 1492 | 59 | |
AK035247.1 | - | 4 - 5 | 653 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |