Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp6r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172894.2 | NP_766482.2 | 4 - 5 | 1033 | 58 | |
GenBank mRNA | AK080891.1 | - | 4 - 5 | 1074 | 58 | |
AK122450.1 | - | 4 - 5 | 1033 | 58 | ||
BC053076.1 | - | 4 - 5 | 1033 | 5698212 | 58 | |
BC141039.1 | - | 3 - 4 | 580 | 9055651 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |