Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199224.1 | NP_001186153.1 | 13 - 14 | 1831 | 111 | |
GenBank mRNA | AK052433.1 | - | 7 - 8 | 1317 | 111 | |
DQ867033.1 | - | 7 - 8 | 1317 | 111 | ||
FN429927.1 | - | 13 - 14 | 1831 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |