Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlgap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145965.1 | NP_001139437.1 | 5 - 6 | 1485 | 103 | |
NM_172910.3 | NP_766498.2 | 6 - 7 | 1584 | 103 | ||
GenBank mRNA | AK032564.1 | - | 6 - 7 | 1585 | 103 | |
AK082945.1 | - | 4 - 5 | 1357 | 103 | ||
AY243847.3 | - | 1 - 2 | 996 | 103 | ||
BC118969.2 | - | 5 - 6 | 1271 | 40097764 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |