Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtmr12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172958.3 | NP_766546.1 | 5 - 6 | 596 | 65 | |
GenBank mRNA | AK030139.1 | - | 5 - 6 | 663 | 65 | |
AK050099.1 | - | 5 - 6 | 589 | 65 | ||
AK122539.1 | - | 5 - 6 | 654 | 65 | ||
BC042573.1 | - | 5 - 6 | 585 | 65 | ||
BC054352.1 | - | 5 - 6 | 596 | 3661020 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |