Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtmr12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172958.3 | NP_766546.1 | 10 - 11 | 1117 | 112 | |
GenBank mRNA | AK030139.1 | - | 10 - 11 | 1184 | 112 | |
AK050099.1 | - | 10 - 11 | 1110 | 112 | ||
AK050236.1 | - | 10 - 11 | 1080 | 112 | ||
AK122539.1 | - | 10 - 11 | 1175 | 112 | ||
BC042573.1 | - | 10 - 11 | 1106 | 112 | ||
BC054352.1 | - | 10 - 11 | 1117 | 3661020 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |