Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adck5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172960.3 | NP_766548.2 | 11 - 12 | 1294 | 54 | |
NR_073123.1 | 11 - 12 | 1289 | 54 | |||
GenBank mRNA | AK039217.1 | - | 11 - 12 | 1290 | 54 | |
AK163557.1 | - | 10 - 11 | 1321 | 54 | ||
BC030881.1 | - | 8 - 9 | 1517 | 54 | ||
BC053696.1 | - | 1 - 2 | 482 | 54 | ||
BC060528.1 | - | 11 - 12 | 1178 | 6414642 | 54 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |