Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phtf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172992.3 | NP_766580.3 | 5 - 6 | 541 | 69 | |
GenBank mRNA | AK035536.1 | - | 5 - 6 | 495 | 69 | |
AK037021.1 | - | 5 - 6 | 428 | 69 | ||
AK042382.1 | - | 5 - 6 | 520 | 69 | ||
AK042790.1 | - | 5 - 6 | 463 | 69 | ||
AK152951.1 | - | 5 - 6 | 494 | 69 | ||
AW060751.1 | - | 1 - 2 | 69 | |||
BC052841.1 | - | 5 - 6 | 454 | 30050876 | 69 | |
CB245749.1 | - | 5 - 6 | 541 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |