Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tango6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173037.1 | NP_766625.1 | 6 - 7 | 1395 | 54 | |
GenBank mRNA | AK047206.1 | - | 1 - 2 | 113 | 54 | |
AK054283.1 | - | 6 - 7 | 1349 | 54 | ||
AK076373.1 | - | 6 - 7 | 1318 | 54 | ||
AK077757.1 | - | 6 - 7 | 1380 | 54 | ||
AK085030.1 | - | 6 - 7 | 1395 | 54 | ||
AK173250.1 | - | 6 - 7 | 1302 | 54 | ||
BC066134.1 | - | 6 - 7 | 1325 | 54 | ||
BC066869.1 | - | 6 - 7 | 1325 | 30106406 | 54 | |
BC069838.1 | - | 6 - 7 | 1325 | 30106422 | 54 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |