Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pmpca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173180.3 | NP_775272.1 | 5 - 6 | 546 | 67 | |
GenBank mRNA | AK004549.1 | - | 5 - 6 | 546 | 67 | |
AK075965.1 | - | 5 - 6 | 547 | 67 | ||
AK077218.1 | - | 5 - 6 | 547 | 67 | ||
AK147110.1 | - | 5 - 6 | 549 | 67 | ||
AK150118.1 | - | 5 - 6 | 547 | 67 | ||
AK158990.1 | - | 5 - 6 | 514 | 67 | ||
AK161291.1 | - | 5 - 6 | 547 | 67 | ||
AK167975.1 | - | 5 - 6 | 545 | 67 | ||
AK169325.1 | - | 5 - 6 | 541 | 67 | ||
AK169342.1 | - | 5 - 6 | 541 | 67 | ||
BC010810.1 | - | 5 - 6 | 531 | 4241697 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |