Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081278.2 | NP_001074747.2 | 7 - 8 | 2367 | 72 | |
GenBank mRNA | AK031120.1 | - | 7 - 8 | 2211 | 72 | |
AK039337.1 | - | 7 - 8 | 2368 | 72 | ||
AK044719.1 | - | 7 - 8 | 2009 | 72 | ||
AK220347.1 | - | 6 - 7 | 774 | 72 | ||
BC042747.1 | - | 3 - 4 | 369 | 72 | ||
BC064433.1 | - | 7 - 8 | 1991 | 72 | ||
BC080762.1 | - | 7 - 8 | 1985 | 72 | ||
BC117869.1 | - | 7 - 8 | 1840 | 40095526 | 72 | |
BC117870.1 | - | 7 - 8 | 1843 | 40095531 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |