Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tph2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173391.3 | NP_775567.2 | 8 - 9 | 1202 | 63 | |
GenBank mRNA | AY090565.1 | - | 8 - 9 | 1202 | 63 | |
BC120514.1 | - | 8 - 9 | 1193 | 40129437 | 63 | |
BC125430.1 | - | 8 - 9 | 1193 | 40129945 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |