Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Haus6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173400.2 | NP_775576.2 | 2 - 3 | 430 | 81 | |
GenBank mRNA | AK040101.1 | - | 2 - 3 | 433 | 81 | |
AK051016.1 | - | 2 - 3 | 450 | 81 | ||
AK077681.1 | - | 2 - 3 | 450 | 81 | ||
BC068237.1 | - | 2 - 3 | 430 | 30550558 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |