Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rccd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173445.4 | NP_775621.2 | 3 - 4 | 1092 | 66 | |
GenBank mRNA | AK030872.1 | - | 3 - 4 | 1063 | 66 | |
AK088662.1 | - | 3 - 4 | 733 | 66 | ||
BC069844.1 | - | 3 - 4 | 1072 | 6487223 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |