Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc24a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175034.3 | NP_778199.2 | 4 - 5 | 536 | 96 | |
GenBank mRNA | AB085629.1 | - | 4 - 5 | 539 | 96 | |
AK089225.1 | - | 4 - 5 | 540 | 96 | ||
AY236527.1 | - | 4 - 5 | 506 | 96 | ||
BC094232.1 | - | 4 - 5 | 496 | 30538386 | 96 | |
BC144866.1 | - | 4 - 5 | 531 | 9053414 | 96 | |
BC145643.1 | - | 4 - 5 | 531 | 40130946 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |