Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Metap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175224.4 | NP_780433.1 | 7 - 8 | 664 | 84 | |
GenBank mRNA | AK077694.1 | - | 7 - 8 | 665 | 84 | |
AK129054.1 | - | 2 - 3 | 2771 | 84 | ||
AK152138.1 | - | 7 - 8 | 683 | 84 | ||
AK153231.1 | - | 7 - 8 | 683 | 84 | ||
AK153944.1 | - | 7 - 8 | 672 | 84 | ||
BC027110.1 | - | 2 - 3 | 204 | 84 | ||
BC096469.1 | - | 7 - 8 | 655 | 3603051 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |