Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coro2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175484.3 | NP_780693.2 | 4 - 5 | 859 | 62 | |
GenBank mRNA | AK047527.1 | - | 4 - 5 | 801 | 62 | |
AK087414.1 | - | 4 - 5 | 860 | 62 | ||
BC062649.1 | - | 4 - 5 | 591 | 6416110 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |