Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fchsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175684.4 | NP_783615.2 | 16 - 17 | 1640 | 79 | |
GenBank mRNA | AK079461.1 | - | 10 - 11 | 1112 | 79 | |
AK154732.1 | - | 16 - 17 | 1675 | 79 | ||
BC052886.1 | - | 16 - 17 | 1671 | 79 | ||
BC057367.1 | - | 16 - 17 | 1640 | 6827858 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |