Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snhg11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175692.3 | NP_783623.2 | 3 - 4 | 579 | 75 | |
GenBank mRNA | AK080745.1 | - | 3 - 4 | 537 | 75 | |
AK157790.1 | - | 4 - 5 | 636 | 75 | ||
BC030414.1 | - | 3 - 4 | 495 | 5366924 | 75 | |
BC049241.1 | - | 493 | 4505955 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |