Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnksr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177751.2 | NP_808419.1 | 9 - 10 | 1224 | 83 | |
GenBank mRNA | BC043093.1 | - | 9 - 10 | 1224 | 6405389 | 83 |
BC060716.1 | - | 9 - 10 | 1224 | 6405389 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |