Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176848.1 | NP_789818.1 | 4 - 5 | 649 | 84 | |
GenBank mRNA | AK140175.1 | - | 4 - 5 | 693 | 84 | |
BC027053.1 | - | 3 - 4 | 602 | 84 | ||
BC046586.1 | - | 4 - 5 | 649 | 6487759 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |