Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024200.4 | NP_077162.2 | 5 - 6 | 753 | 69 | |
GenBank mRNA | AK018181.1 | - | 5 - 6 | 750 | 69 | |
AK046313.1 | - | 5 - 6 | 706 | 69 | ||
AK141611.1 | - | 5 - 6 | 754 | 69 | ||
AK220169.1 | - | 5 - 6 | 741 | 69 | ||
AY174062.1 | - | 4 - 5 | 882 | 69 | ||
BC056641.1 | - | 5 - 6 | 712 | 5097345 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |