Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnmt3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003960.4 | NP_001003960.2 | 6 - 7 | 894 | 83 | |
NM_001003961.4 | NP_001003961.2 | 6 - 7 | 894 | 83 | ||
NM_001003963.4 | NP_001003963.2 | 6 - 7 | 894 | 83 | ||
NM_001122997.2 | NP_001116469.1 | 5 - 6 | 779 | 83 | ||
NM_001271744.1 | NP_001258673.1 | 5 - 6 | 779 | 83 | ||
NM_001271745.1 | NP_001258674.1 | 5 - 6 | 779 | 83 | ||
NM_001271746.1 | NP_001258675.1 | 5 - 6 | 779 | 83 | ||
NM_001271747.1 | NP_001258676.1 | 5 - 6 | 779 | 83 | ||
NM_010068.5 | NP_034198.3 | 6 - 7 | 894 | 83 | ||
GenBank mRNA | AF068626.2 | - | 6 - 7 | 749 | 83 | |
AF068627.2 | - | 6 - 7 | 749 | 83 | ||
AF068628.2 | - | 6 - 7 | 749 | 83 | ||
AF151969.1 | - | 6 - 7 | 892 | 83 | ||
AF151970.1 | - | 6 - 7 | 892 | 83 | ||
AF151971.1 | - | 6 - 7 | 892 | 83 | ||
AF151972.1 | - | 6 - 7 | 892 | 83 | ||
AF151973.1 | - | 5 - 6 | 777 | 83 | ||
AF151974.1 | - | 5 - 6 | 777 | 83 | ||
AF151975.1 | - | 5 - 6 | 777 | 83 | ||
AF151976.1 | - | 5 - 6 | 777 | 83 | ||
AY078427.1 | - | 6 - 7 | 749 | 83 | ||
BC105677.1 | - | 5 - 6 | 611 | 40048574 | 83 | |
BC105922.1 | - | 5 - 6 | 611 | 40048584 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |