Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gart | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010256.2 | NP_034386.2 | 19 - 20 | 2719 | 140 | |
GenBank mRNA | AK146355.1 | - | 19 - 20 | 2704 | 140 | |
AK146478.1 | - | 19 - 20 | 2661 | 140 | ||
AK168458.1 | - | 19 - 20 | 140 | |||
AK168501.1 | - | 19 - 20 | 2682 | 140 | ||
AK168724.1 | - | 19 - 20 | 2694 | 140 | ||
AK168796.1 | - | 19 - 20 | 2694 | 140 | ||
AK168864.1 | - | 19 - 20 | 2670 | 140 | ||
AK168876.1 | - | 19 - 20 | 2671 | 140 | ||
BC070465.1 | - | 19 - 20 | 2666 | 30611835 | 140 | |
U01024.1 | - | 18 - 19 | 2624 | 140 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |