Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pon1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011134.3 | NP_035264.2 | 1 - 2 | 262 | 73 | |
GenBank mRNA | AK050119.1 | - | 1 - 2 | 80 | 73 | |
BC012706.1 | - | 1 - 2 | 135 | 4158951 | 73 | |
U32684.1 | - | 1 - 2 | 144 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |