Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rph3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB112924.1 | - | 11 - 12 | 1127 | 146 | |
BC042585.1 | - | 11 - 12 | 1294 | 5364086 | 146 | |
BC050883.1 | - | 11 - 12 | 1261 | 5698459 | 146 | |
D29965.1 | - | 11 - 12 | 1104 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |