Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013599.3 | NP_038627.1 | 9 - 10 | 1701 | 72 | |
GenBank mRNA | AK004651.1 | - | 9 - 10 | 1682 | 72 | |
AK142787.1 | - | 9 - 10 | 1683 | 72 | ||
AK159292.1 | - | 9 - 10 | 1683 | 72 | ||
AK161176.1 | - | 9 - 10 | 1682 | 72 | ||
AY622969.1 | - | 1 - 2 | 84 | 72 | ||
BC046991.1 | - | 9 - 10 | 1669 | 6309245 | 72 | |
D12712.1 | - | 9 - 10 | 1690 | 72 | ||
S67830.1 | - | 9 - 10 | 1710 | 72 | ||
X72795.1 | - | 7 - 8 | 1268 | 72 | ||
Z27231.1 | - | 9 - 10 | 1682 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |