Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024434.6 | NP_077754.3 | 4 - 5 | 470 | 72 | |
GenBank mRNA | AF334160.1 | - | 4 - 5 | 431 | 72 | |
AK002819.1 | - | 4 - 5 | 406 | 72 | ||
AK005334.1 | - | 4 - 5 | 383 | 72 | ||
AK008600.1 | - | 4 - 5 | 357 | 72 | ||
AK010384.1 | - | 4 - 5 | 356 | 72 | ||
AK010502.1 | - | 4 - 5 | 423 | 72 | ||
AK011604.1 | - | 4 - 5 | 356 | 72 | ||
AK089023.1 | - | 4 - 5 | 437 | 72 | ||
AK166958.1 | - | 4 - 5 | 354 | 72 | ||
AK169032.1 | - | 4 - 5 | 356 | 72 | ||
BC016536.1 | - | 4 - 5 | 470 | 3488469 | 72 | |
CT010237.1 | - | 4 - 5 | 384 | 72 | ||
CT010301.1 | - | 4 - 5 | 384 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |