Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080728.2 | NP_542766.1 | 11 - 12 | 1035 | 138 | |
GenBank mRNA | AK040973.1 | - | 11 - 12 | 1037 | 138 | |
AK147031.1 | - | 11 - 12 | 1036 | 138 | ||
AK157742.1 | - | 11 - 12 | 1036 | 138 | ||
AK169198.1 | - | 11 - 12 | 1035 | 138 | ||
AK169371.1 | - | 11 - 12 | 1035 | 138 | ||
AY056464.1 | - | 11 - 12 | 1035 | 138 | ||
BC121789.1 | - | 10 - 11 | 991 | 40099448 | 138 | |
BC158018.1 | - | 1112 | 9007324 | 138 | ||
BC158070.1 | - | 11 - 12 | 1290 | 9007368 | 138 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |