Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dis3l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172157.1 | NP_001165628.1 | 19 - 20 | 2493 | 76 | |
NM_153530.2 | NP_705758.1 | 18 - 19 | 2451 | 76 | ||
GenBank mRNA | AK029652.1 | - | 19 - 20 | 2508 | 76 | |
AK031180.1 | - | 19 - 20 | 2476 | 76 | ||
BC036177.1 | - | 18 - 19 | 2412 | 5004341 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |