Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dirc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153550.3 | NP_705778.1 | 5 - 6 | 1059 | 99 | |
GenBank mRNA | AK034214.1 | - | 5 - 6 | 1059 | 99 | |
AK172331.1 | - | 5 - 6 | 1074 | 99 | ||
BC037460.1 | - | 5 - 6 | 1033 | 4190539 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |