Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbln7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024237.4 | NP_077199.2 | 1 - 2 | 213 | 80 | |
GenBank mRNA | AK005465.1 | - | 1 - 2 | 168 | 80 | |
BB867118.1 | - | 1 - 2 | 227 | 80 | ||
BC095941.1 | - | 1 - 2 | 145 | 4167912 | 80 | |
CK023294.1 | - | 1 - 2 | 155 | 80 | ||
EF668007.1 | - | 1 - 2 | 72 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |