Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027332.3 | NP_081608.1 | 3 - 4 | 396 | 124 | |
NM_027371.3 | NP_081647.2 | 3 - 4 | 396 | 124 | ||
GenBank mRNA | AK008988.1 | - | 3 - 4 | 385 | 124 | |
AK010079.1 | - | 3 - 4 | 393 | 124 | ||
AK162073.1 | - | 3 - 4 | 399 | 124 | ||
AK171079.1 | - | 3 - 4 | 385 | 124 | ||
BC056165.1 | - | 3 - 4 | 347 | 5694049 | 124 | |
BC089369.1 | - | 3 - 4 | 381 | 30550759 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |