Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134077.4 | NP_598838.3 | 10 - 11 | 1998 | 80 | |
GenBank mRNA | AK049083.1 | - | 9 - 10 | 1453 | 80 | |
AK051686.1 | - | 10 - 11 | 1974 | 80 | ||
AK139077.1 | - | 10 - 11 | 1999 | 80 | ||
AK172039.1 | - | 10 - 11 | 1977 | 80 | ||
BC049360.1 | - | 10 - 11 | 1975 | 80 | ||
BC066051.1 | - | 10 - 11 | 1950 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |