Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnot10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153585.5 | NP_705813.2 | 18 - 19 | 2341 | 69 | |
GenBank mRNA | AK029103.1 | - | 18 - 19 | 2216 | 69 | |
AK149944.1 | - | 18 - 19 | 2171 | 69 | ||
AK171701.1 | - | 15 - 16 | 3261 | 69 | ||
BC025554.1 | - | 18 - 19 | 2255 | 5321869 | 69 | |
BC036178.1 | - | 15 - 16 | 1797 | 69 | ||
BC039183.1 | - | 18 - 19 | 2315 | 2631793 | 69 | |
BC047204.1 | - | 2 - 3 | 353 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |