Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173788.3 | NP_776149.1 | 8 - 9 | 1806 | 81 | |
GenBank mRNA | AK158195.1 | - | 3 - 4 | 357 | 81 | |
BC037458.1 | - | 3 - 4 | 262 | 81 | ||
BC042470.1 | - | 8 - 9 | 1806 | 5137261 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |