Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Micall2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174850.3 | NP_777275.2 | 10 - 11 | 2617 | 78 | |
GenBank mRNA | AB182579.1 | - | 10 - 11 | 2468 | 78 | |
AK044547.1 | - | 10 - 11 | 2469 | 78 | ||
AK151086.1 | - | 10 - 11 | 2616 | 78 | ||
AK164043.1 | - | 10 - 11 | 2588 | 78 | ||
AK165558.1 | - | 10 - 11 | 2467 | 78 | ||
AK220308.1 | - | 10 - 11 | 2394 | 78 | ||
BC043300.1 | - | 10 - 11 | 2431 | 5025620 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |