Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam73a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174868.4 | NP_777357.2 | 10 - 11 | 1465 | 65 | |
GenBank mRNA | AK044739.1 | - | 10 - 11 | 1441 | 65 | |
AK083909.1 | - | 9 - 10 | 1296 | 65 | ||
AK085017.1 | - | 10 - 11 | 1465 | 65 | ||
AK143366.1 | - | 10 - 11 | 1465 | 65 | ||
AK166051.1 | - | 10 - 11 | 1418 | 65 | ||
BC098205.1 | - | 10 - 11 | 1448 | 6409928 | 65 | |
BC138328.1 | - | 10 - 11 | 1412 | 8861348 | 65 | |
BC138329.1 | - | 10 - 11 | 1412 | 8861349 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |