Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175173.3 | NP_780382.2 | 22 - 23 | 2260 | 61 | |
GenBank mRNA | AK040117.1 | - | 7 - 8 | 700 | 61 | |
AK077050.1 | - | 22 - 23 | 2269 | 61 | ||
AV277048.2 | - | 2 - 3 | 178 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |