Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atxn7l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289545.1 | NP_001276474.1 | 5 - 6 | 851 | 62 | |
NM_175183.5 | NP_780392.1 | 6 - 7 | 1030 | 62 | ||
GenBank mRNA | AK046849.1 | - | 6 - 7 | 1438 | 62 | |
BC088996.1 | - | 4 - 5 | 625 | 62 | ||
BC094547.1 | - | 5 - 6 | 786 | 62 | ||
BC137599.1 | - | 5 - 6 | 772 | 8860618 | 62 | |
BC137600.1 | - | 5 - 6 | 799 | 8860619 | 62 | |
BC144756.1 | - | 5 - 6 | 779 | 9053304 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |