Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sppl2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175195.3 | NP_780404.2 | 12 - 13 | 1325 | 68 | |
GenBank mRNA | AK046804.1 | - | 12 - 13 | 1344 | 68 | |
AK148109.1 | - | 12 - 13 | 1292 | 68 | ||
AK166364.1 | - | 12 - 13 | 1276 | 68 | ||
AK170375.1 | - | 12 - 13 | 1291 | 68 | ||
BC034206.1 | - | 9 - 10 | 880 | 68 | ||
BC052094.1 | - | 10 - 11 | 999 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |