Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rapgef6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252494.1 | NP_001239423.1 | 10 - 11 | 1301 | 116 | |
NM_001252496.1 | NP_001239425.1 | 10 - 11 | 1301 | 116 | ||
NM_001252497.1 | NP_001239426.1 | 5 - 6 | 1559 | 116 | ||
NM_001252498.1 | NP_001239427.1 | 5 - 6 | 1559 | 116 | ||
NM_175258.4 | NP_780467.2 | 10 - 11 | 1301 | 116 | ||
GenBank mRNA | AK045540.1 | - | 10 - 11 | 1243 | 116 | |
AK049228.1 | - | 10 - 11 | 1257 | 116 | ||
AK049669.1 | - | 5 - 6 | 1559 | 117 | ||
AK164343.1 | - | 5 - 6 | 1532 | 116 | ||
AK167055.1 | - | 10 - 11 | 1302 | 116 | ||
BC141182.1 | - | 10 - 11 | 1200 | 9055794 | 116 | |
BC145503.1 | - | 10 - 11 | 1200 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |